EMAIL THIS PAGE TO A FRIEND
Biochemistry
Radical peregrinations catalyzed by coenzyme B12-dependent enzymes.
PMID 11371179
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0884
Coenzyme B12, ≥97.0%
C72H100CoN18O17P