EMAIL THIS PAGE TO A FRIEND
Diabetes
IGF-1 overexpression inhibits the development of diabetic cardiomyopathy and angiotensin II-mediated oxidative stress
PMID 11375343
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-284
Anti-Nitrotyrosine Antibody, clone, Upstate®, from rabbit