EMAIL THIS PAGE TO A FRIEND
Gastroenterology
Targeting cyclooxygenase 2 and HER-2/neu pathways inhibits colorectal carcinoma growth.
PMID 11375952
Abstract
The cyclooxygenase 2 (COX-2) and ErbB/HER pathways are important modulators of cancer cell growth. We sought to determine the effects of treatment with a specific COX-2 inhibitor and/or a monoclonal antibody against the ErbB receptor subtype HER-2/neu on carcinoma cell growth. A cell-proliferation assay was used to determine the response of HCA-7 cells to the HER-3/HER-4 ligand heregulin beta-1 (HRGbeta-1). Both in vitro and in vivo assays were used to determine the effects of the selective COX-2 inhibitor, celecoxib, and/or an anti-HER-2/neu monoclonal antibody (either Herceptin [Genetech Inc., S. San Francisco, CA] or 2C4) on cell growth. HCA-7 cells express HER-2/neu messenger RNA and protein, and exposure of these cells to HRGbeta-1 results in a significant stimulation of cell growth. Celecoxib or Herceptin inhibits HCA-7 cell growth in vitro and in vivo. Combination therapy with celecoxib plus Herceptin or celecoxib plus 2C4 resulted in additive effects that resulted in almost complete inhibition of tumor growth. Combined treatment with COX-2 and HER-2/neu inhibitors more effectively reduces colorectal carcinoma growth than either agent alone. Therefore, targeting of both the COX-2 and ErbB signaling pathways may represent a novel approach for the treatment and/or prevention of colorectal cancer in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
HCA-7 Colony 29, 02091238