EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Sorbitol-fermenting Shiga toxin-producing Escherichia coli O157:H(-) strains: epidemiology, phenotypic and molecular characteristics, and microbiological diagnosis.
PMID 11376032