EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
LC determination of the enantiomeric purity of levamisole using stationary phase with bonded naphthylethylcarbamoylated-beta-cyclodextrin.
PMID 11377020
Abstract
A direct enantioselective high-performance liquid chromatography was employed successfully for determination of the enantiomeric purity of levamisole. The elaborated method used S-naphthylethylcarbamoylated beta-cyclodextrin stationary phase in reversed-phase mode. The optimized mobile phase composition was acetonitrile-0.5% triethylammonium acetate buffer, pH 5.0 (2:8, v/v). Linearity, precision, accuracy, and the quantitation limit were determined. The method proved to be capable of determining 0.05% (w/w) of dexamisole (the enantiomeric impurity) contrary to the pharmacopoeial optical rotation measurement, in which only amounts of dexamisole higher than 2.2% (w/w) caused the test to fail. The enantiomeric purity of three different levamisole substances and levamisole tablets was assessed with the use of the method. The content of dexamisole impurity was found to be in the range 0.66-1.60% (w/w).