EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Structure-activity relationships of phenylcyclohexene and biphenyl antitubulin compounds against plant and mammalian cells.
PMID 11378362
Abstract
Phenylcyclohexenes (PCHs) [e.g., trans-4-nitro-5-(2,3,4-trimethoxyphenyl)cyclohexene, 2d] were found to bind weakly to the colchicine site of bovine tubulin, but are the first mimics of colchicine found to have high activity towards plant cells. Structure-activity relationships for PCHs and biphenyl AC-ring analogues of colchicine (e.g., 2,3,4,4'-tetramethoxy-2'-methyl-1,1'-biphenyl, 3e) are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P22303
1-Phenyl-1-cyclohexene, 95%
C12H14