EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Occupational erythema-multiforme-like dermatitis from sensitization to costus resinoid, followed by flare-up and systemic contact dermatitis from beta-cyclocostunolide in a chemistry student.
PMID 11379984
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W233609 Costus oil, natural, FG