EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Use of SE-30 as a stationary phase for the gas-liquid chromatography of drugs.
PMID 1138012
Abstract
The dimethyl silicone elastomer SE-30 has been chosen as the preferred liquid phase for the gas-liquid chromatographic analysis of drugs, and retention index data have been compiled for 480 drugs and commonly occurring chemicals such as plasticisers. The inter-laboratory variation in measurement of retention indices has been measured for three drugs in eleven laboratories and the standard deviations were between 20 and 15 retention index units.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21099-U GC Stationary Phase, phase SE-30, bottle of 10 g