EMAIL THIS PAGE TO A FRIEND
British dental journal
The influence of two different dental local anaesthetic solutions on the haemodynamic responses of children undergoing restorative dentistry: a randomised, single-blind, split-mouth study.
PMID 11384023
Abstract
This investigation was designed to study the haemodynamic effects of two different local anaesthetic solutions during restorative dental treatment in children. A randomised, single-blind, split-mouth cross-over design was employed using children undergoing bilaterally similar restorative treatments over two visits. The study was performed in a dental hospital paediatric dentistry department. Ten children participated. At one visit the local anaesthetic was 2% lidocaine (lignocaine) with 1:80,000 epinephrine (adrenaline); at the other the anaesthetic was 3% prilocaine with 0.03IU/ml felypressin. Local anaesthetic was administered at a dose of 0.5 ml/10 kg body weight. Blood pressure and heart rate were measured before and during treatment with an automatic blood pressure recorder. Data were analysed by ANOVA and Student's paired t test. Significant differences between treatments in diastolic blood pressure (F = 2.37; P = 0.05) and heart rate (F = 2.98; P< 0.02) were noted. The heart rate increased ten minutes following the injection of the epinephrine-containing solution. The diastolic blood pressure fell 20 minutes after injection of lidocaine with epinephrine. The choice of local anaesthetic solution influences the haemodynamic response during restorative treatment in children.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000415 Felypressin, European Pharmacopoeia (EP) Reference Standard
C46H65N13O11S2