EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Quantitative estimation of renal clearance of N-acetylprocainamide in rats with various experimental acute renal failure.
PMID 11384853
Abstract
The dosage regimen of a drug eliminated predominantly through the kidney need to be adjusted for the patients with renal disease. The objective of the present study was to establish a quantitative approach to precisely predicting the renal clearances of basic drugs using N-1-methylnicotinamide (NMN). A variety of experimental acute renal failure (ARF) in rats were prepared and N-acetylprocainamide (NAPA) was used as a model drug. The renal clearances of NAPA were significantly decreased in rats with ARF, resulting in significantly increased plasma concentrations. Remarkable reduction in clearance ratios (CL(ratio)) was observed, indicating that the impairment in tubular and glomerular function did not proceed in a parallel manner. The renal clearance of NAPA (CL(rNAPA)) was better predicted from the renal clearance of NMN (CL(rNMN)) than from GFR. A mathematical equation was also constructed to estimate the CL(rNMN) from the NMN plasma concentration. Therefore, the renal clearance of basic drugs excreted predominantly from the kidney can be easily and more accurately estimated based on the concentrations of endogenous NMN to provide a precise dosage regimen for patients with renal failure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269476
N-Acetylprocainamide, ≥99%
C15H23N3O2
A5513
N-Acetylprocainamide hydrochloride, ≥99% (HPLC), powder
C15H23N3O2 · HCl