EMAIL THIS PAGE TO A FRIEND
The Analyst
Determination of 3,5-dinitro-o-toluamide in feedstuffs and pre-mixes.
PMID 1138581
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
524417
3,5-Dinitro-o-toluamide, 98%
C8H7N3O5