EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Turnover of oxidatively damaged nuclear proteins in BV-2 microglial cells is linked to their activation state by poly-ADP-ribose polymerase.
PMID 11387257
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6510
N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin, ≥90% (HPLC)
C40H53N5O10