EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Biosynthesis of 2-aminobenzaldehyde in flowers of Robinia pseudoacacia and Philadelphus coronarius.
PMID 11393514
Abstract
Feeding experiments with 13C- and fluorine-labelled precursors were performed to reveal the biosynthesis of 2-aminobenzaldehyde in flowers of Robinia pseudoacacia and Philadelphus coronarius. The results are in agreement with the transformation of anthranilic acid to indole followed by oxidative ring opening and hydrolysis of the resulting N-formyl-2-aminobenzaldehyde. This route differs from that observed in Hebeloma sacchariolens (Basidiomycetes) in which anthranilic acid is directly reduced to 2-aminobenzaldehyde.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9628
2-Aminobenzaldehyde, ≥98%
C7H7NO