EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Transcription factor Yin Yang 1 stimulates poly(ADP-ribosyl)ation and DNA repair.
PMID 11394900
Abstract
Poly(ADP-ribose) polymerase 1 (PARP-1) is a nuclear enzyme that catalyzes the synthesis of ADP-ribose polymers from NAD(+). The function of PARP-1 is related to important nuclear processes including DNA repair and transcription. Previous studies demonstrated a specific physical interaction between PARP-1 and the transcription factor Yin Yang 1 (YY1) in vitro. In this study, a functional relationship between both proteins in response to genotoxic treatment of cells is presented. The interaction of YY1 with PARP-1 greatly stimulates the enzymatic activity of PARP-1. Consistent with this, the overexpression of YY1 in HeLa cells resulted in an enhanced synthesis of poly(ADP-ribose) and an acceleration of DNA repair in response to a treatment with methyl-N'-nitro-N'-nitrosoguanidine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0996 PARP-1 human, recombinant, expressed in E. coli