EMAIL THIS PAGE TO A FRIEND
Current opinion in obstetrics & gynecology
Endometrial effects of etonogestrel (Implanon) contraceptive implant.
PMID 11396660
Abstract
The subdermal contraceptive implant etonogestrel (Implanon; NV Organon International, Oss, The Netherlands) exerts complex effects on the endometrium. These include direct effects on the endometrium through endometrial progestin target sites, and indirect effects through suppression of the hypothalamic-pituitary-ovarian axis. The resulting effects are categorized by alterations in endometrial histology, endometrial thickness, dysmenorrhoea and menstrual bleeding pattern. The exact mechanism of action of progestins on the endometrium has not been determined. The contraceptive effect in Implanon users is mainly due to inhibition of ovulation. Current research is concentrating on the potential of the progestin implant to modify endometrial vascular, angiogenic, steroid receptor and proto-oncogene function. These processes may be involved in the causation of progestin-induced breakthrough bleeding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0356
Etonogestrel, ≥98% (HPLC)
C22H28O2