EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Rapid homogeneous-phase Sonogashira coupling reactions using controlled microwave heating.
PMID 11397149
Abstract
A microwave-enhanced, rapid and efficient homogeneous-phase version of the Sonogashira reaction is presented. It has been applied to the coupling of aryl iodides, bromides, triflates, and aryl chloride, as well as pyridine and thiophene derivatives with trimethylsilylacetylene. Excellent yields (80-95%) for substrates containing a large variety of substituents in different positions are obtained in 5-25 min.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196150
2-Iodothiophene, 98%
C4H3IS