EMAIL THIS PAGE TO A FRIEND
Oral surgery, oral medicine, oral pathology, oral radiology, and endodontics
Evaluation of oral antimicrobial agent levels in tooth extraction sites.
PMID 11402275
Abstract
The purpose of this study was to evaluate various oral antimicrobial agent levels in tooth extraction sites. The concentration of dental alveolar blood in extraction wounds after the oral administration of talampicillin (500 mg), cefaclor (500 mg), cefteram pivoxil (200 mg), cefuroxime axetil (250 mg), cefdinir (200 mg), and ofloxacin (100 mg) was determined in 338 patients and was assessed on the basis of its antimicrobial activity against Streptococcus isolated in odontogenic infections. The percentage of patients whose concentrations exceeded the minimum inhibitory concentration for 90% of Streptococcus was 62.5% to 100% for talampicillin at 30 to 360 minutes, 0% to 12.5% for cefaclor at 30 to 360 minutes, 18.2% to 100% for cefteram pivoxil at 30 to 480 minutes, 50% to 100% for cefuroxime axetil at 30 to 480 minutes, 0% to 50% for cefdinir at 16 to 290 minutes, and 0% to 40% for ofloxacin at 30 to 480 minutes. These results indicate that talampicillin, cefteram pivoxil, and cefuroxime axetil have minimum inhibitory concentration levels for 90% of Streptococcus in tooth sockets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0010000 Talampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C24H23N3O6S · HCl