EMAIL THIS PAGE TO A FRIEND
Acta veterinaria Hungarica
Hepatic biotransformation profiles of sulphamonomethoxine in food-producing animals and rats in vitro.
PMID 11402713
Abstract
Hydroxylation and acetylation of sulphamonomethoxine (SMM) and deacetylation of N4-acetyl SMM (N4-AcSMM) were estimated in liver post-mitochondrial supernatants (S-9) from laying hens, female cattle, swine and rats. The formation of hydroxylated SMM, 2,6-dihydroxy SMM (2,6-diOH-SMM), was found only with hen S-9s. N4-acetylation rate of SMM was the highest in pig S-9s, followed by rat, then hen or cow S-9s. All S-9s from the four species deacetylated N4-AcSMM. In hen S-9s, the rate of 2,6-dihydroxylation was higher during incubation at 41 degrees C than at 37 degrees C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0508
Sulfamonomethoxine
C11H12N4O3S
32091
Sulfamonomethoxine, VETRANAL, analytical standard
C11H12N4O3S