EMAIL THIS PAGE TO A FRIEND
Organic letters
Efficient scavenging of Ph3P and Ph3P=O with high-loading merrifield resin.
PMID 11405732
Abstract
[see reaction]. A simple, highly effective method for removing triphenylphosphine and/or triphenylphosphine oxide from reaction mixtures is described. Commercially available high-loading chloromethylated polystyrene 1, modified in situ with NaI, acts as a scavenger resin. Several coupling reactions catalyzed by Pd(0) or Ni(0) which require the removal of triphenylphosphine are tested.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T84603
Triphenylphosphine oxide, 98%
C18H15OP