EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Anaerobic biodegradation of DDT residues (DDT, DDD, and DDE) in estuarine sediment.
PMID 11411851
Abstract
The potential for anaerobic biodegradation of 1,1,1-trichloro-2,2-bischlorophenylethane (DDT), 1,1-dichloro-2,2,-bischlorophenylethane (DDD), and dichlorodiphenylchloroethylene (DDE) in anoxic sediment slurries collected from the Keelung River was investigated in this study. o,p'- and p,p'-DDT were dechlorinated to o,p'- and p,p'-DDD, respectively, and then transformed to other compound(s). 1-Chloro-2,2-bis (p-chlorophenyl) ethylene (DDMU) and trace amount of dichlorobenzophenone (DBP) were detected in sediment slurries amended with p,p'-DDT or p,p'-DDD. DDMU was also detected in sediment slurries amended with p,p'-DDE. The relative transformation rates for both o,p'- and p,p'-isomers of DDT, DDD, and DDE were DDT>DDD>DDE. Re-addition of DDT, DDD, or DDE to the sediment slurries after initial removal enhanced the respective dechlorination rates. The transformation rates of the p,p'-isomers of both DDT and DDD were faster than those of the respective o,p'-isomers. p,p'-DDT dechlorination in the p,p'-DDT-adapted sediment slurries were inhibited by the addition of molybdate, or molybdate plus sulfate, but not inhibited by the addition of sulfate. Addition of bromoethane-sulfonic acid (BESA) slightly inhibited p,p'-DDT dechlorination. Non-adapted sediment slurries lost the ability to dechlorinate pentachlorophenol during adaptation to p,p'-DDT. p,p'-DDD was the major transformation product of p, p'-DDT in 3,4,4',5-tetrachlorobiphenyl-adapted sediment slurries, which suggested that the microbial community in the 3,4,4',5-CB-adapted sediment was unable to remove chlorine from the aromatic rings of p,p'-DDT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
137502
Sodium 2-bromoethanesulfonate, 98%
C2H4BrNaO3S