EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
New probes of the agonist binding site of metabotropic glutamate receptors.
PMID 11412983
Abstract
The (2S,4R)- and (2S,4S)-4-hydroxyglutamates activate cloned mGlu(1a), mGlu(2), and mGlu(8a) receptors with different potencies. Best results were obtained with the (2S,4S) isomer being almost as potent as glutamate on mGlu(1a)R and mGlu(8a)R. Data are interpreted on the basis of the binding site model and X-ray structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76157
(4R)-4-Hydroxy-L-glutamic acid, ≥98.0% (TLC)
C5H9NO5