EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Chronic beryllium disease: T cell recognition of a metal presented by HLA-DP.
PMID 11414740
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14270
Beryllium sulfate tetrahydrate, purum p.a., ≥99.0% (T)
BeO4S · 4H2O