EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Synergy between liquid chromatographic-pulsed amperometric detection and capillary-gas chromatographic methods for the detection of juice adulteration.
PMID 11417660
Abstract
The use of liquid chromatography linked with pulsed amperometric detection, and capillary-gas chromatography to detect the adulteration of fruit juices is discussed. These are 2 complementary approaches which can be used to detect the addition of sugar syrups to juices at low levels.