EMAIL THIS PAGE TO A FRIEND
FEBS letters
Human leukocyte glycosylasparaginase: cell-to-cell transfer and properties in correction of aspartylglycosaminuria.
PMID 11418116
Abstract
Aspartylglycosaminuria (AGU), a severe lysosomal storage disease, is caused by the deficiency of the lysosomal enzyme, glycosylasparaginase (GA), and accumulation of aspartylglucosamine (GlcNAc-Asn) in tissues. Here we show that human leukocyte glycosylasparaginase can correct the metabolic defect in Epstein-Barr virus (EBV)-transformed AGU lymphocytes rapidly and effectively by mannose-6-phosphate receptor-mediated endocytosis or by contact-mediated cell-to-cell transfer from normal EBV-transformed lymphocytes, and that 2-7% of normal activity is sufficient to correct the GlcNAc-Asn metabolism in the cells. Cell-to-cell contact is obligatory for the transfer of GA since normal transformed lymphocytes do not excrete GA into extracellular medium. The combined evidence indicates that cell-to-cell transfer of GA plays a main role in enzyme replacement therapy of AGU by normal lymphocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6681
2-Acetamido-1-N-(β-L-aspartyl)-2-deoxy-β-D-glucopyranosylamine, ≥98%
C12H21N3O8