EMAIL THIS PAGE TO A FRIEND
Oncogene
New agents in cancer clinical trials.
PMID 11426656
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0040
Lometrexol hydrate, ≥95% (HPLC)
C21H25N5O6 · xH2O