EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Cathepsin B, L, and S cleave and inactivate secretory leucoprotease inhibitor.
PMID 11435427
Abstract
A number of serine proteases, matrix metalloproteases, and cysteine proteases were evaluated for their ability to cleave and inactivate the antiprotease, secretory leucoprotease inhibitor (SLPI). None of the serine proteases or the matrix metalloproteases examined cleaved the SLPI protein. However, incubation with cathepsins B, L, and S resulted in the cleavage and inactivation of SLPI. All three cathepsins initially cleaved SLPI between residues Thr(67) and Tyr(68). The proteolytic cleavage of SLPI by all three cathepsins resulted in the loss of the active site of SLPI and the inactivation of SLPI anti-neutrophil elastase capacity. Cleavage and inactivation were catalytic with respect to the cathepsins, so that the majority of a 400-fold excess of SLPI was inactivated within 15 min by cathepsins L and S. Analysis of epithelial lining fluid samples from individuals with emphysema indicated the presence of cleaved SLPI in these samples whereas only intact SLPI was observed in control epithelial lining fluid samples. Active cathepsin L was shown to be present in emphysema epithelial lining fluid and inhibition of this protease prevented the cleavage of recombinant SLPI added to emphysema epithelial lining fluid. Taken together with previous data that demonstrates that cathepsin L inactivates alpha(1)-antitrypsin, these findings indicate the involvement of cathepsins in the diminution of the lung antiprotease screen possibly leading to lung destruction in emphysema.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6854 Cathepsin L from human liver, ≥0.5 units/mg protein, solution