EMAIL THIS PAGE TO A FRIEND
Protein expression and purification
Characterization of Itk tyrosine kinase: contribution of noncatalytic domains to enzymatic activity.
PMID 11437596
Abstract
Itk is a Tec family tyrosine kinase found in T cells that is activated upon ligation of the T cell receptor (TCR/CD3), CD2, or CD28. Itk contains five domains in addition to the catalytic domain: pleckstrin homology, Tec homology which contains a proline-rich region, Src homology 3, and Src homology 2. To provide a basis for understanding the contribution of these various domains to catalysis, recombinant Itk was purified and its substrate specificity determined by steady-state kinetic methods. Measurements of the rates of phosphorylation of various protein substrates, including Src associated in mitosis 68K protein (SAM68), CD28, linker for activation of T cells, and CD3 zeta, at a fixed concentration indicated that SAM68 was phosphorylated most rapidly. Wild-type Itk and three Itk mutants were characterized by comparing their activity (k(cat)) using the SAM68 substrate. A deletion mutant removing the pleckstrin homology domain and part of the Tec homology domain (Itk(Delta152)) had approximately 10-fold less activity than wild type, a mutant with an altered proline-rich domain (P158A,P159A) had a more dramatic 100-fold loss of activity, and the catalytic domain alone was essentially inactive. Itk(Delta152) had K(m) values for ATP and SAM68 nearly identical to those of the wild-type enzyme, while Itk(P158A,P159A) had approximately 3-fold higher K(m) values for each substrate. SAM68 phosphorylation by the wild-type and mutant enzymes in the presence of several tyrosine kinase inhibitors were compared using a homogeneous time-resolved fluorescence assay. Both the Itk(Delta152) deletion mutant and the Itk(P158A,P159A) mutant had IC(50) values similar to those of the wild-type enzyme for staurosporine, PP1, and damnacanthal. These comparisons, taken together with the similar K(m) values for ATP and SAM68 substrate between the wild-type and the mutant enzymes, indicate that the amino acids in the N-terminal 152 residues and proline-rich domains enhance catalysis by affecting turnover rate rather than substrate binding.