EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
The hydrolytic and transferase action of alternanase on oligosaccharides.
PMID 11438094
Abstract
Alternanase is an enzyme which endo-hydrolytically cleaves the alpha-(1-->3), alpha-(1-->6)-linked D-glucan, alternan. The main products are isomaltose, alpha-D-Glcp-(1-->3)-alpha-D-Glcp-(1-->6)-D-Glc and the cyclic tetrasaccharide cyclo[-->6)-alpha-D-Glcp-(1-->3)-alpha-D-Glcp-(1-->6)-alpha-D-Glcp-(1-->3)-alpha-D-Glcp-(1-->]. It is also capable of acting on oligosaccharide substrates. The cyclic tetrasaccharide is slowly hydrolyzed to isomaltose. Panose and the trisaccharide alpha-D-Glcp-(1-->6)-alpha-D-Glcp-(1-->3)-D-Glc both undergo transglycosylation reactions to give rise to the cyclic tetrasaccharide plus D-glucose, with panose being converted at a much faster rate. The tetrasaccharide alpha-D-Glcp-(1-->3)-alpha-D-Glcp-(1-->6)-alpha-D-Glcp-(1-->4)-D-Glc is hydrolyzed to D-glucose plus the trisaccharide alpha-D-Glcp-(1-->3)-alpha-D-Glcp-(1-->6)-D-Glc. Alternanase does not act on isomaltotriose, theanderose (6(Glc)-O-alpha-D-Glcp sucrose), or alpha-D-Glcp-(1-->6)-alpha-D-Glcp-(1-->6)-alpha-D-Glcp-(1-->4)-alpha-D-Glc. The enzyme releases 4-nitrophenol from 4-nitrophenyl alpha-isomaltoside, but not from 4-nitrophenyl alpha-D-glucopyranoside, 4-nitrophenyl alpha-isomaltotrioside, or 4-nitrophenyl alpha-isomaltotetraoside.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2407
D-Panose, ≥97%
C18H32O16