EMAIL THIS PAGE TO A FRIEND
Chemosphere
Photoinductive properties of soil humic acids and their fractions obtained by tandem size exclusion chromatography-polyacrylamide gel electrophoresis.
PMID 11444301
Abstract
Humic acids (HAs) from three soils of different origin (Chernozem, Ferralsol and Ranker) have been fractionated by coupling size exclusion chromatography (SEC) and polyacrylamide gel electrophoresis (PAGE) on three fractions (fractions A, B, C + D) with different molecular sizes (MSs) and exactly defined electrophoretic mobility (EM). Fractions identically marked had similar EM and MS, independently of HA sources. The photoinductive properties of the whole HAs and their fractions were compared by studying the photoinduced transformation of fenuron at 365 nm. High MS fractions A and B appeared to exhibit poor photoinductive activities compared to the whole HAs, whereas low MS fraction C + D in Chernozem and Ranker were more efficient than the whole HAs. A fourth intermediary fraction containing a mixture of fractions B and C + D with small amount of D was shown to photoinduce poorly the transformation of fenuron. It was therefore concluded that the molecules capable of photoinducing the transformation of fenuron were mainly contained in fraction D. Fluorescence properties of Chernozem HA and its fractions have been tested. Fraction C + D exhibited a very similar fluorescence emission spectrum in comparison with the whole HA and in contrast, the fractions A and B emitted very weakly.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45494
Fenuron, PESTANAL®, analytical standard
C9H12N2O