EMAIL THIS PAGE TO A FRIEND
Fitoterapia
The major flavonoid of Dodonaea angustifolia.
PMID 11449522
Abstract
The major leaf flavonoid of Dodonaea angustifolia, an important South African traditional medicine, has been identified as 5,7,4'-trihydroxy-3,6-dimethoxyflavone (1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00102
4′,5,7-Trihydroxy-3,6-dimethoxyflavone, ≥90% (LC/MS-ELSD)
C17H14O7