EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Pyrrolizidine alkaloids from Senecio lorenthii.
PMID 11449528
Abstract
The isolation and NMR spectra of otosenine (1) and seneciphylline (2) from Senecio lorenthii are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73913
Seneciphylline, analytical standard
C18H23NO5