EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
Interaction mechanism between indoxyl sulfate, a typical uremic toxin bound to site II, and ligands bound to site I of human serum albumin.
PMID 11451040
Abstract
The study was performed for clarifying the mechanism of interaction between indoxyl sulfate (IS), a typical uremic toxin bound to site II, and site I-ligands when bound to human serum albumin (HSA). The effect of the N to B transition on the interactions was also examined. Quantitative investigation of the relations between ligands bound to HSA was performed by equilibrium dialysis, and the binding data were analyzed on the basis of a theoretical model for simultaneous binding of two ligands. The high-affinity binding constants for the site I-ligands warfarin (WF) and dansyl-L-asparagine (DNSA) increased with increasing pH, whereas those for the site II-ligands IS and dansylsarcosine (DNSS) were hardly affected by pH. Mutual displacement experiments showed that even though IS binds to site II it influenced binding of DNSA at the azapropazone binding area in site I. By contrast, it is unlikely that IS affects the WF binding area of site I. Furthermore, pH-profiles showed that the interaction between IS and DNSA was very sensitive to the N to B transition: "competitive-like" strong allosteric regulation was observed for binding of the two ligands to the N conformer (pH 6.5), whereas in the B conformation (pH 8.5) binding of these molecules was nearly "independent". The present data provide useful information for elucidating a potential mechanism of interaction between drugs and endogenous substances including uremic toxins.