EMAIL THIS PAGE TO A FRIEND
Arzneimittel-Forschung
Etofenamate levels in human serum and synovial fluid following iontophoresis.
PMID 11455681
Abstract
The absorption of etofenamate (CAS 30544-47-9, Rheumon gel) by iontophoresis in 11 patients with low back pain and in 13 patients with synovitis of the knee was evaluated. During the 5-day treatment period, the test gel in a quantity corresponding to 100 mg etofenamate was applied to affected body regions every day by 20-min iontophoresis sessions. Two hours after the fifth application, the concentration of etofenamate in serum and synovial fluid (in patients who had knee joint iontophoresis) were measured by HPLC. Iontophoresis of etofenamate into the lumbar region as well as to the knee joint resulted in consistent serum levels: 219 +/- 136.3 micrograms/l and 191 +/- 84.6 micrograms/l, respectively. In patients with synovitis of the knee, the synovial level of etofenamate (368 +/- 109.2 micrograms/l) was almost twice as high than the serum concentration. The authors conclude that with topical application of etofenamate by iontophoresis the drug appears not only in the serum but also--with higher levels--in the synovial fluid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2499900 Etofenamate, European Pharmacopoeia (EP) Reference Standard
C18H18F3NO4