EMAIL THIS PAGE TO A FRIEND
Acta veterinaria Scandinavica
Eradication of lice in cattle.
PMID 11455904
Abstract
The purpose of this field study was to develop and evaluate eradication as a strategy to control lice in cattle. Thirty-three herds of cattle were selected and observed during a period of two and a half years. Before eradication, biting lice (Damalinia bovis) were present in 94% of the herds and 27% of the animals. Sucking lice (Linognathus vituli) were present in 42% of the herds and 5% of the animals. These levels were very similar to those reported from other countries in Northern Europe. The eradication strategy was successful in 28 of 33 herds, but lice were still present in 5 herds 3 to 6 months after treatment. Biting lice were present in all these 5 herds, sucking lice were present in 3 herds. During the next 12 months, nine of the 28 herds were reinfected with lice. Six herds were reinfected with just biting lice, 2 herds with just sucking lice and one herd was reinfected with both. There was no significant difference between the 2 louse species regarding the risk of unsuccessful eradication or reinfection. The only significant risk factor for reinfection was either purchase of livestock or use of common pasture, combined with failure in pre-treatment of newly introduced animals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46417
Flumethrin, PESTANAL®, analytical standard
C28H22Cl2FNO3