EMAIL THIS PAGE TO A FRIEND
Water research
The variation on the mutagenicity of CNP during anaerobic biodegradation.
PMID 11456156
Abstract
The mutagenicity of water, including herbicide CNP, and its time-variation during anaerobic biodegradation were studied through Ames assay using strains with or without. S9 mix: TA98, TA 100, YG1021, YG1024, YG1026, and YG1029. The bacteria, for the anaerobic biodegradation, was obtained from a paddy field, and preincubated for a month. The CNP was decomposed in an anaerobic culture inoculated with the bacteria, and finally yielded CNP-amino as one of the CNP metabolites. About 16% of the initial CNP was transformed into CNP-amino by the 14th day. The mutagenicities to TA98. YG1024, and YG1029 strains with S9 mix increased with cultivating time, the latter two showed the strongest sensitivity to CNP-amino. The contribution of CNP to the mutagenicity decreased as the chemical decomposed, while the contribution of CNP-amino increased. However, the increased mutagenicity was not limited to the contribution of CNP-amino. but also to the contribution of other metabolites. The contributions of other CNP metabolites were 67% of total mutagenicity to the TA98 strain and 30% to the YG1029 strain. These unknown mutagenic metabolites were the indirect frameshift mutagens which did not have nitro- and amino-substituents, and the indirect base-pair mutagens which might possibly have some amino-substituents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04287
Chlornitrofen, analytical standard
C12H6Cl3N1O3