EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
The asymmetric [3 + 2] cycloaddition reaction of chiral alkenyl Fischer carbene complexes with imines: synthesis of optically pure 2,5-disubstituted-3-pyrrolidinones.
PMID 11459508
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388122
Tin(II) trifluoromethanesulfonate, 97%
C2F6O6S2Sn