EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Novel tricyclic benzothiazolo[2,3-c]thiadiazine antagonists of the platelet ADP receptor (P2Y(12)).
PMID 11459636
Abstract
Novel non-nucleoside tricyclic platelet ADP receptor (P2Y(12)) antagonists have been discovered that bind reversibly and with high affinity to the platelet receptor. Condensation of various 2-aminobenzothiazoles with chlorosulfonylacetyl chloride affords these novel tricyclic heterocycles, which are novel and unpredicted products of this reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
259772
Chlorosulfonylacetyl chloride, 95%
C2H2Cl2O3S