EMAIL THIS PAGE TO A FRIEND
Marine environmental research
Increased potential for NAD(P)H-dependent reactive oxygen species production of hepatic subcellular fractions of fish species with in vivo exposure to contaminants.
PMID 11460751
Abstract
The present study investigated the proposed involvement of contaminant-stimulated reactive oxygen species (ROS) production in disease processes in fish. NAD(P)H-dependent ROS production of subcellular fractions was determined by the iron/EDTA-mediated oxidation of 2-keto-4-methiolbutyric acid. Hepatic cytosolic NADPH-dependent and microsomal NAD(P)H-dependent ROS production were increased 51-160% (P < 0.05) in rainbow trout (Oncorhynchus mykiss) 15 weeks after a single i.p. injection of polychlorobiphenyl (PCB) (100 mg Clophen A50 kg-1 wet wt.). Hepatic microsomal NADH-dependent ROS production was 114% higher in perch (Perca fluviatilis) from PCB-contaminated Lake Järnsjön compared to clean Lake Vänern, Sweden. Hepatic mitochondrial NADH-dependent, cytosolic NADH-dependent and microsomal NADPH-dependent ROS production were variously elevated up to 160% in flounder (Platichthys flesus) at various sites along two pollution transects near to the ports of Rotterdam and Amsterdam, Netherlands. Overall the data indicate increased potential for ROS production in liver of fish exposed to field pollution, and support the hypothesis of oxidative stress as a mechanism of contaminant-mediated disease in fish.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K6000
α-Keto-γ-(methylthio)butyric acid sodium salt, ≥97%
C5H7NaO3S