EMAIL THIS PAGE TO A FRIEND
European journal of immunology
ULBP1, 2, 3: novel MHC class I-related molecules that bind to human cytomegalovirus glycoprotein UL16, activate NK cells.
PMID 11465099
Abstract
New members of the extended MHC class I-like family were identified based on their ability to bind human cytomegalovirus glycoprotein UL16 and/or their mutual homology. Soluble UL16 binding proteins (ULBP) competed with each other for binding to NK cells. Treatment of human and mouse NK cells with ULBP led to increased production of cytokines/chemokines, proliferation, cytotoxic activity and up-regulation of activation-associated surface molecules. The presence of ULBP during the stimulation phase of the CTL assay caused increased cytotoxic activity. Addition of soluble recombinant UL16 protein inhibited the biological activities mediated by ULBP, suggesting the existence of a novel mechanism utilized by CMV to evade elimination by the host immune system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3501063
Anti-ULBP1 antibody produced in rabbit, affinity isolated antibody