EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Cyclic AMP-dependent protein kinase isoenzymes in human myeloid leukemia (HL60) and breast tumor (MCF-7) cells.
PMID 11469804
Abstract
Combinations of retinoic acid (RA) and cAMP mediate many biological responses in a large variety of cell types. While the basis for the apparent synergistic effects of RA and cAMP are not clearly defined, it is likely that activation of PKA by cAMP is involved. However, literature reports concerning the identity of PKA isoforms in HL60 and MCF-7 cells are conflicting. The purpose of the present investigation is to identify PKA isoforms in HL60 and MCF-7 cells. Utilization of high-performance anion-exchange liquid chromatography, immunoblotting, and 8-azido-cAMP photoaffinity binding resulted in the finding that HL60 cells contain PKA types I alpha and II alpha, while MCF-7 cells contain PKA types I alpha, II alpha, and II beta. PKA type I alpha in both HL60 and MCF-7 cells eluted from columns as two well-separated peaks. One peak eluted at a low salt concentration in agreement with previous reports. The second HL60 PKA type I alpha peak eluted at a salt concentration intermediate between that eluting the first peak and that eluting PKA type II alpha and contained approximately 62% of the total RI alpha protein. However, the second MCF-7 PKA type I alpha peak contained approximately 66% of the total RI alpha protein and co-eluted with PKA types II alpha and II beta. This "contamination" of PKA type II fractions with PKA type I has led, in some cases, to interpretations that may need reevaluation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1262
8-Azidoadenosine 3′:5′-cyclic monophosphate, ~95%
C10H11N8O6P