EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Ion solvation and association in LiClO4/sulfolane solution: a vibrational spectroscopic and molecular orbital study.
PMID 11471707
Abstract
Solvation interaction and ion association in solutions of lithium perchlorate/sulfolane have been studied by using infrared and Raman spectra as a function of concentration of lithium perchlorate. The band changes of antisymmetric OSO stretch, antisymmetric CSC stretch, -SO2 wag and twist suggest that there is an interaction between Li+ and sulfolane molecules, and the site of solvation is the oxygen atom of -SO2 group. The molecular orbital calculation supports this suggestion. On the other hand, the apparent solvation number was calculated, and the band fitting for the ClO4- band reveals the presence of contact ion pair, solvent separated ion pair and free ClO4- anion in the concentrated solutions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88965
Sulfolane, analytical standard
C4H8O2S
T22209
Sulfolane, 99%
C4H8O2S