EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
Direct visualization of protein interactions in plant cells.
PMID 11479572
Abstract
The protein NPR1/NIM1 is required for the induction of systemic acquired resistance (SAR) in plants and has been shown to interact with members of the TGA/OBF family of basic leucine zipper (bZIP) transcription factors. However, to date, there is no method available to monitor such interactions in plant cells. We report here an in vivo protein fragment complementation assay (PCA), based on association of reconstituted murine dihydrofolate reductase (mDHFR) with a fluorescent probe to detect protein-protein interaction in planta. We demonstrate that the interaction between Arabidopsis NPR1/NIM1 and the bZIP factor TGA2 is induced by the regulators of SAR, salicylic acid (SA), and its analog 2,6-dichloroisonicotinic acid (INA) with distinct species-specific responses. Furthermore, the induced interaction is localized predominantly in the nucleus. Protein fragment complementation assays could be of value to agricultural research by providing a system for high-throughput biochemical pathway mapping and for screening of small molecules that modulate protein interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
456543
2,6-Dichloropyridine-4-carboxylic acid, 98%
C6H3Cl2NO2