EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Calcium sensors as new therapeutic targets for airway hyperresponsiveness and asthma.
PMID 11481245
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V5640 Calcium-Like Peptide 3