EMAIL THIS PAGE TO A FRIEND
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina
The influence of the analogue 4-10 corticotropin derivative on the general structure of the rabbits behaviour in stress.
PMID 11482057
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0401
Adrenocorticotropic Hormone Fragment 4-10 human, rat, ≥97% (HPLC)
C44H59N13O10S