EMAIL THIS PAGE TO A FRIEND
Organic letters
Mild and selective sodium azide mediated cleavage of p-nitrobenzoic esters.
PMID 11483039
Abstract
[reaction: see text] A mild and selective cleavage of p-nitrobenzoic esters by sodium azide in methanol is reported. This new methodology is mild enough for use with acid- or base-sensitive compounds. No elimination byproducts are formed. Fmoc- and trifluoroacetyl-amino protecting groups, benzyl esters, and ethyl esters remain unaffected. Less reactive compounds are discussed in terms of steric factors, and yields are increased by altering the azide solvation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72910
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)
C7H5NO4
461091
4-Nitrobenzoic acid, 98%
C7H5NO4