EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of beta-lactones by the regioselective, cobalt and Lewis acid catalyzed carbonylation of simple and functionalized epoxides.
PMID 11485465
Abstract
The PPNCo(CO)(4) and BF(3) x Et(2)O catalyzed carbonylation of simple and functionalized epoxides in DME gives the corresponding beta-lactones regioselectively in good to high yields. The carbonylation occurred selectively at the unsubstituted C-O bond of the epoxide ring, and this reaction tolerates various functional groups such as alkenyl, halide, hydroxy, and alkyl ether.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219126
β-Butyrolactone, 98%
C4H6O2