EMAIL THIS PAGE TO A FRIEND
Biodegradation
Transformation of 2,4,6-trichlorophenol by the white rot fungi Panus tigrinus and Coriolus versicolor.
PMID 11487063
Abstract
The toxicity of thirteen isomers of mono-, di-, tri- and pentachlorophenols was tested in potato-dextrose agar cultures of the white rot fungi Panus tigrinus and Coriolus versicolor. 2,4,6-Trichlorophenol (2,4,6-TCP) was chosen for further study of its toxicity and transformation in liquid cultures of these fungi. Two schemes of 2,4,6-TCP addition were tested to minimize its toxic effect to fungal cultures: stepwise addition from the moment of inoculation and single addition after five days of growth. In both cases the ligninolytic enzyme systems of both fungi were found to be responsible for 2,4,6-TCP transformation. 2,6-Dichloro-1,4-hydroquinol and 2,6-dichloro-1,4-benzoquinone were found as products of primary oxidation of 2,4,6-TCP by intact fungal cultures and purified ligninolytic enzymes, Mn-peroxidases and laccases of both fungi. However, primary attack of 2,4,6-TCP in P. tigrinus culture was conducted mainly by Mn-peroxidase, while in C. versicolor it was catalyzed predominantly by laccase, suggesting a different mode of regulation of these enzymes in the two fungi.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
431982
2,6-Dichloro-1,4-benzoquinone, 98%
C6H2Cl2O2