EMAIL THIS PAGE TO A FRIEND
Allergy
Contact cross-sensitization among quinolines.
PMID 11488685
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130532
5,7-Dichloro-8-hydroxy-2-methylquinoline, 98%
C10H7Cl2NO