EMAIL THIS PAGE TO A FRIEND
Journal of endodontics
Cytotoxicity of halothane on human gingival fibroblast cultures in vitro.
PMID 11491643
Abstract
Recently halothane has been reported to be the most suitable alternative to chloroform in dissolving gutta-percha. Periapical tissue toxicity of halothane is not completely known. In this study gutta-percha dissolved by halothane was evaluated with the almar blue dye assay using human gingival fibroblast cultures. The cytotoxic effects of halothane on human gingival fibroblasts depended on the exposure dose, frequency, and duration. A reduced concentration and smaller amount of gutta-percha solvents may minimize the cytotoxic effects on host tissues.