Description
Molecular Formula
Add to Cart
CKOZFN1002 CompoZr® Knockout ZFN Kit, human ABL1 (NM_005157)
CKOZFN1003 CompoZr® Knockout ZFN Kit, human ADAM17 (NM_003183)
CKOZFN1147 CompoZr® Knockout ZFN Kit, human ADRB3 (NM_000025)
CKOZFN1148 CompoZr® Knockout ZFN Kit, human ADRB3 (NM_000025)
CKOZFN1153 CompoZr® Knockout ZFN Kit, human AGTRAP (NM_020350)
CKOZFN1152 CompoZr® Knockout ZFN Kit, human AGTRAP (NM_020350)
CKOZFN1004 CompoZr® Knockout ZFN Kit, human AR (NM_000044)
CKOZFN1136 CompoZr® Knockout ZFN Kit, human AURKB (NM_004217)
CKOZFN1079 CompoZr® Knockout ZFN Kit, human AVPR2 (NM_000054)
CKOZFN1078 CompoZr® Knockout ZFN Kit, human AVPR2 (NM_000054)
CKOZFN1005 CompoZr® Knockout ZFN Kit, human BACE1 (NM_012104)
CKOZFN1006 CompoZr® Knockout ZFN Kit, human BAX (NM_004324)
CKOZFN1007 CompoZr® Knockout ZFN Kit, human BCL2 (NM_000633)
CKOZFN1132 CompoZr® Knockout ZFN Kit, human CACNA2D4 (NM_172364)
CKOZFN1008 CompoZr® Knockout ZFN Kit, human CBLB (NM_170662)
CKOZFN1164 CompoZr® Knockout ZFN Kit, human CCR5 (NM_000579)
CKOZFN1120 CompoZr® Knockout ZFN Kit, human CD19 (NM_001770)
CKOZFN1121 CompoZr® Knockout ZFN Kit, human CD33 (NM_001772)
CKOZFN1113 CompoZr® Knockout ZFN Kit, human CD40 (NM_001250)
CKOZFN1119 CompoZr® Knockout ZFN Kit, human CD8A (NM_001768)
CKOZFN1118 CompoZr® Knockout ZFN Kit, human CD8A (NM_001768)
CKOZFN1131 CompoZr® Knockout ZFN Kit, human CDCP1 (NM_022842)
CKOZFN1009 CompoZr® Knockout ZFN Kit, human CDH1 (NM_004360)
CKOZFN1010 CompoZr® Knockout ZFN Kit, human CDKN1B (NM_004064)
CKOZFN1080 CompoZr® Knockout ZFN Kit, human CETP (NM_000078)
CKOZFN1011 CompoZr® Knockout ZFN Kit, human CFLAR (NM_003879)
CKOZFN1012 CompoZr® Knockout ZFN Kit, human CHEK1 (NM_001274)
CKOZFN1013 CompoZr® Knockout ZFN Kit, human CLTB (NM_001834)
CKOZFN1150 CompoZr® Knockout ZFN Kit, human CNR1 (NM_016083)
CKOZFN1149 CompoZr® Knockout ZFN Kit, human CNR1 (NM_016083)